Product Details

SNP ID
rs140472566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25675258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGGCTTCTCCTTTGAGTGTATC[C/T]GCCAGTGGTGTCTGGTGAGACTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF337 PubMed Links

Gene Details

Gene
ZNF337
Gene Name
zinc finger protein 337
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290261.1 1901 Missense Mutation CAG,CGG Q677R NP_001277190.1
NM_015655.3 1901 Missense Mutation CAG,CGG Q677R NP_056470.1
XM_005260702.3 1901 Missense Mutation CAG,CGG Q645R XP_005260759.1
XM_006723558.3 1901 Missense Mutation CAG,CGG Q677R XP_006723621.1
XM_011529219.2 1901 Missense Mutation CAG,CGG Q677R XP_011527521.1
XM_017027802.1 1901 Missense Mutation CAG,CGG Q645R XP_016883291.1
XM_017027803.1 1901 Missense Mutation CAG,CGG Q645R XP_016883292.1
XM_017027804.1 1901 Missense Mutation CAG,CGG Q612R XP_016883293.1
Gene
ZNF337-AS1
Gene Name
ZNF337 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details