Product Details

SNP ID
rs140489990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33668433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTGCAGTTTGAATCTCCCCAA[C/T]CACTGCCAGTTCAGAGAATGCCAGT
Phenotype
MIM: 189971 MIM: 612478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTL10 PubMed Links

Gene Details

Gene
ACTL10
Gene Name
actin like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024675.1 1936 UTR 3 NP_001019846.1
Gene
C20orf144
Gene Name
chromosome 20 open reading frame 144
There are no transcripts associated with this gene.

Gene
E2F1
Gene Name
E2F transcription factor 1
There are no transcripts associated with this gene.

Gene
NECAB3
Gene Name
N-terminal EF-hand calcium binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031231.3 1936 Intron NP_112508.3
NM_031232.3 1936 Intron NP_112509.3
XM_005260510.1 1936 Intron XP_005260567.1
XM_011528991.1 1936 Intron XP_011527293.1
XM_011528992.2 1936 Intron XP_011527294.1
XM_017028015.1 1936 Intron XP_016883504.1
XM_017028016.1 1936 Intron XP_016883505.1

View Full Product Details