Product Details

SNP ID
rs141149688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4687720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCAGTTTCACTGCTGTAATGA[C/T]GAGGATCTTCATGGCCGCGTTATTT
Phenotype
MIM: 176640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRNP PubMed Links

Gene Details

Gene
PRNP
Gene Name
prion protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000311.3 Intron NP_000302.1
NM_001080121.1 Intron NP_001073590.1
NM_001080122.1 Intron NP_001073591.1
NM_001080123.1 Intron NP_001073592.1
NM_001271561.1 Intron NP_001258490.1
NM_183079.2 Intron NP_898902.1

View Full Product Details