Product Details

SNP ID
rs141396923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32197617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGGTTCCGCAGATGGTCCTTG[C/T]GGTGAAACATCTTATCACAGTACAT
Phenotype
MIM: 604866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLAGL2 PubMed Links

Gene Details

Gene
PLAGL2
Gene Name
PLAG1 like zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002657.3 1388 Intron NP_002648.1
XM_005260436.3 1388 Missense Mutation CAC,CGC H109R XP_005260493.1
XM_011528863.2 1388 Missense Mutation CAC,CGC H109R XP_011527165.1
XM_011528864.2 1388 Missense Mutation CAC,CGC H109R XP_011527166.1
Gene
TSPY26P
Gene Name
testis specific protein, Y-linked 26, pseudogene
There are no transcripts associated with this gene.

View Full Product Details