Product Details

SNP ID
rs141483339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37522377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAGGTGTTCCTGGAATGCTGCA[C/T]TTACTGGGTAGGATTCGCTTTTCGA
Phenotype
MIM: 613110 MIM: 603106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLCAP PubMed Links

Gene Details

Gene
BLCAP
Gene Name
bladder cancer associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167820.1 218 Intron NP_001161292.1
NM_001167821.1 218 Intron NP_001161293.1
NM_001167822.2 218 Intron NP_001161294.1
NM_001167823.1 218 Intron NP_001161295.1
NM_001317074.1 218 Intron NP_001304003.1
NM_001317075.1 218 Intron NP_001304004.1
NM_006698.3 218 Intron NP_006689.1
XM_017027599.1 218 Intron XP_016883088.1
Gene
NNAT
Gene Name
neuronatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322802.1 218 Missense Mutation ACT,ATT T31I NP_001309731.1
NM_005386.3 218 Missense Mutation ACT,ATT T31I NP_005377.1
NM_181689.2 218 Intron NP_859017.1

View Full Product Details