Product Details

SNP ID
rs142111439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36615270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGAGTGAGGGGAAGGTGAGGC[A/G]CGGTGAGATGTACAGCCAGCCATTG
Phenotype
MIM: 606577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf24 PubMed Links

Gene Details

Gene
C20orf24
Gene Name
chromosome 20 open reading frame 24
There are no transcripts associated with this gene.

Gene
SLA2
Gene Name
Src like adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032214.3 901 Missense Mutation CGC,TGC R163C NP_115590.1
NM_175077.2 901 Missense Mutation CGC,TGC R163C NP_778252.1
XM_017028098.1 901 Missense Mutation CGC,TGC R163C XP_016883587.1
Gene
TGIF2-C20orf24
Gene Name
TGIF2-C20orf24 readthrough
There are no transcripts associated with this gene.

View Full Product Details