Product Details

SNP ID
rs142166964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37522703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGCAGAAGCTGGCATACACG[A/G]TGTCGCGGACCGGGCGGCAGGTGTT
Phenotype
MIM: 613110 MIM: 603106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLCAP PubMed Links

Gene Details

Gene
BLCAP
Gene Name
bladder cancer associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167820.1 235 Intron NP_001161292.1
NM_001167821.1 235 Intron NP_001161293.1
NM_001167822.2 235 Intron NP_001161294.1
NM_001167823.1 235 Intron NP_001161295.1
NM_001317074.1 235 Intron NP_001304003.1
NM_001317075.1 235 Intron NP_001304004.1
NM_006698.3 235 Intron NP_006689.1
XM_017027599.1 235 Intron XP_016883088.1
Gene
NNAT
Gene Name
neuronatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322802.1 235 Missense Mutation GAT,GGT D62G NP_001309731.1
NM_005386.3 235 Missense Mutation ATG,GTG M64V NP_005377.1
NM_181689.2 235 Missense Mutation ATG,GTG M37V NP_859017.1

View Full Product Details