Product Details
- SNP ID
-
rs142896442
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:37184276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACCAAGCATGACGTGGAGAGAC[G/T]AAAAGCCTCGCTGGATCGCCCTTTC
- Phenotype
-
MIM: 180490
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MROH8
PubMed Links
Gene Details
- Gene
- MROH8
- Gene Name
- maestro heat like repeat family member 8
There are no transcripts associated with this gene.
- Gene
- RPN2
- Gene Name
- ribophorin II
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135771.2 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_001129243.1 |
NM_001324299.1 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_001311228.1 |
NM_001324301.1 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_001311230.1 |
NM_001324302.1 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_001311231.1 |
NM_001324303.1 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_001311232.1 |
NM_001324304.1 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_001311233.1 |
NM_001324305.1 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_001311234.1 |
NM_001324306.1 |
421 |
UTR 5 |
|
|
NP_001311235.1 |
NM_002951.4 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
NP_002942.2 |
XM_006723851.2 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
XP_006723914.1 |
XM_006723852.2 |
421 |
Missense Mutation |
CGA,CTA |
R37L |
XP_006723915.1 |
View Full Product Details