Product Details

SNP ID
rs143582801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33238110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGGGCATTCTGGAAAACCTTCC[A/G]CTCCTGGACATCCTGAAGCCTGGAG
Phenotype
MIM: 607412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPIFA1 PubMed Links

Gene Details

Gene
BPIFA1
Gene Name
BPI fold containing family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243193.1 286 Silent Mutation CCA,CCG P72P NP_001230122.1
NM_016583.3 286 Silent Mutation CCA,CCG P72P NP_057667.1
NM_130852.2 286 Silent Mutation CCA,CCG P72P NP_570913.1

View Full Product Details