Product Details
- SNP ID
-
rs144649777
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:34715352 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGATTTTCTTCGCTTGGATTGCAC[C/T]GCACTGGTTATGTCTGTGGGGGAAA
- Phenotype
-
MIM: 605299
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NCOA6
PubMed Links
Gene Details
- Gene
- NCOA6
- Gene Name
- nuclear receptor coactivator 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242539.2 |
6490 |
Silent Mutation |
GCA,GCG |
A1061A |
NP_001229468.1 |
NM_001318240.1 |
6490 |
Silent Mutation |
GCA,GCG |
A2054A |
NP_001305169.1 |
NM_014071.4 |
6490 |
Silent Mutation |
GCA,GCG |
A2054A |
NP_054790.2 |
XM_006723755.3 |
6490 |
Intron |
|
|
XP_006723818.1 |
XM_011528721.2 |
6490 |
Silent Mutation |
GCA,GCG |
A2059A |
XP_011527023.1 |
XM_011528726.2 |
6490 |
Silent Mutation |
GCA,GCG |
A2048A |
XP_011527028.1 |
XM_011528727.2 |
6490 |
Silent Mutation |
GCA,GCG |
A2047A |
XP_011527029.1 |
XM_017027739.1 |
6490 |
Silent Mutation |
GCA,GCG |
A2078A |
XP_016883228.1 |
XM_017027740.1 |
6490 |
Silent Mutation |
GCA,GCG |
A2078A |
XP_016883229.1 |
XM_017027741.1 |
6490 |
Silent Mutation |
GCA,GCG |
A2077A |
XP_016883230.1 |
XM_017027742.1 |
6490 |
Silent Mutation |
GCA,GCG |
A2073A |
XP_016883231.1 |
XM_017027743.1 |
6490 |
Missense Mutation |
AGT,GGT |
S2029G |
XP_016883232.1 |
XM_017027744.1 |
6490 |
Silent Mutation |
GCA,GCG |
A2042A |
XP_016883233.1 |
XM_017027745.1 |
6490 |
Silent Mutation |
GCA,GCG |
A2030A |
XP_016883234.1 |
XM_017027746.1 |
6490 |
Intron |
|
|
XP_016883235.1 |
XM_017027747.1 |
6490 |
Silent Mutation |
GCA,GCG |
A1898A |
XP_016883236.1 |
XM_017027748.1 |
6490 |
UTR 3 |
|
|
XP_016883237.1 |
XM_017027749.1 |
6490 |
Intron |
|
|
XP_016883238.1 |
XM_017027750.1 |
6490 |
Silent Mutation |
GCA,GCG |
A1085A |
XP_016883239.1 |
XM_017027751.1 |
6490 |
Silent Mutation |
GCA,GCG |
A1061A |
XP_016883240.1 |
- Gene
- TP53INP2
- Gene Name
- tumor protein p53 inducible nuclear protein 2
There are no transcripts associated with this gene.
View Full Product Details