Product Details

SNP ID
rs145787306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33238120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGAAAACCTTCCGCTCCTGGAC[A/G]TCCTGAAGCCTGGAGGAGGTACTTC
Phenotype
MIM: 607412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPIFA1 PubMed Links

Gene Details

Gene
BPIFA1
Gene Name
BPI fold containing family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243193.1 296 Missense Mutation ATC,GTC I76V NP_001230122.1
NM_016583.3 296 Missense Mutation ATC,GTC I76V NP_057667.1
NM_130852.2 296 Missense Mutation ATC,GTC I76V NP_570913.1

View Full Product Details