Product Details
- SNP ID
-
rs146180205
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63861568 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCGTCATCTGCTTCGTGAAGGCG[C/T]TGGTGCGCGTGTTCAAGATCTACCT
- Phenotype
-
MIM: 603747
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD16B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2281534] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABHD16B
- Gene Name
- abhydrolase domain containing 16B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080622.3 |
356 |
Silent Mutation |
CTG,TTG |
L10L |
NP_542189.1 |
- Gene
- TPD52L2
- Gene Name
- tumor protein D52 like 2
There are no transcripts associated with this gene.
View Full Product Details