Product Details

SNP ID
rs146267300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34929576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGGCCTGTACCATTTCCTCCC[C/T]ATATAGGTTGTTACCTGCAATGAAA
Phenotype
MIM: 605832 MIM: 601002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSS2 PubMed Links

Gene Details

Gene
ACSS2
Gene Name
acyl-CoA synthetase short-chain family member 2
There are no transcripts associated with this gene.

Gene
GSS
Gene Name
glutathione synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000178.3 1225 Missense Mutation AGG,GGG R376G NP_000169.1
NM_001322494.1 1225 Missense Mutation AGG,GGG R376G NP_001309423.1
NM_001322495.1 1225 Missense Mutation AGG,GGG R376G NP_001309424.1

View Full Product Details