Product Details

SNP ID
rs146420040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35176182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATCCACAGTTCCTCTGACCATC[C/T]GCTGCTTCCTGGGCTGTGAGCTGCC
Phenotype
MIM: 600646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROCR PubMed Links

Gene Details

Gene
PROCR
Gene Name
protein C receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006404.4 555 Missense Mutation CGC,TGC R113C NP_006395.2
XM_011528496.1 555 Missense Mutation CGC,TGC R113C XP_011526798.1

View Full Product Details