Product Details

SNP ID
rs146547099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63540883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGTGGCGAAGGAGGGCCGTTCCT[C/T]GGGGCTGCTCCTCCAGCACTCCAGC
Phenotype
MIM: 602004
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTK6 PubMed Links

Gene Details

Gene
PTK6
Gene Name
protein tyrosine kinase 6
There are no transcripts associated with this gene.

Gene
SRMS
Gene Name
src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080823.3 1443 Missense Mutation AAG,GAG K468E NP_543013.1

View Full Product Details