Product Details

SNP ID
rs147407921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43514626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTCGCAATCCTCAGACCCTCCC[A/G]CGCTCCAGTTCCGGATAAGCGAGTA
Phenotype
MIM: 608802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
L3MBTL1 PubMed Links

Gene Details

Gene
L3MBTL1
Gene Name
l(3)mbt-like 1 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015478.6 191 Missense Mutation ACG,GCG T28A NP_056293.4
NM_032107.4 191 Intron NP_115479.4

View Full Product Details