Product Details

SNP ID
rs147514649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35542527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCCCCAGGTGCATCCTGAGCT[C/T]TACGTGGACAAGTCGCGGGGAGATA
Phenotype
MIM: 616971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERGIC3 PubMed Links

Gene Details

Gene
ERGIC3
Gene Name
ERGIC and golgi 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015966.2 243 Silent Mutation CTC,CTT L58L NP_057050.1
NM_198398.1 243 Silent Mutation CTC,CTT L58L NP_938408.1

View Full Product Details