Product Details

SNP ID
rs147571833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57329909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGCCCGAGGCCTCGTTCTTCGA[A/C]GTTTTGGACCGACACAGGGAGTCCC
Phenotype
MIM: 605114
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR4325 PubMed Links

Gene Details

Gene
MIR4325
Gene Name
microRNA 4325
There are no transcripts associated with this gene.

Gene
SPO11
Gene Name
SPO11, initiator of meiotic double stranded breaks
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012444.2 135 Missense Mutation GAA,GAC E14D NP_036576.1
NM_198265.1 135 Missense Mutation GAA,GAC E14D NP_937998.1
XM_005260379.3 135 Missense Mutation GAA,GAC E14D XP_005260436.1
XM_005260380.3 135 Missense Mutation GAA,GAC E14D XP_005260437.1
XM_005260381.3 135 Missense Mutation GAA,GAC E14D XP_005260438.1
XM_005260382.4 135 Missense Mutation GAA,GAC E14D XP_005260439.1
XM_011528756.2 135 Missense Mutation GAA,GAC E14D XP_011527058.1
XM_011528757.2 135 Intron XP_011527059.1

View Full Product Details