Product Details
- SNP ID
-
rs147671569
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1181276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCATAATCCACCGGGCGGCTCA[C/T]GGGCTCCAGGGCTGGCCCCTCCTCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PSMF1
PubMed Links
Gene Details
- Gene
- PSMF1
- Gene Name
- proteasome inhibitor subunit 1
There are no transcripts associated with this gene.
- Gene
- TMEM74B
- Gene Name
- transmembrane protein 74B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001304748.1 |
1196 |
Missense Mutation |
ATG,GTG |
M115V |
NP_001291677.1 |
NM_001304749.1 |
1196 |
Missense Mutation |
ATG,GTG |
M91V |
NP_001291678.1 |
NM_018354.2 |
1196 |
Missense Mutation |
ATG,GTG |
M115V |
NP_060824.1 |
XM_011529281.2 |
1196 |
Missense Mutation |
ATG,GTG |
M172V |
XP_011527583.1 |
XM_011529282.2 |
1196 |
Missense Mutation |
ATG,GTG |
M115V |
XP_011527584.1 |
XM_011529283.2 |
1196 |
Missense Mutation |
ATG,GTG |
M115V |
XP_011527585.1 |
XM_011529284.2 |
1196 |
Missense Mutation |
ATG,GTG |
M115V |
XP_011527586.1 |
XM_011529285.2 |
1196 |
Missense Mutation |
ATG,GTG |
M91V |
XP_011527587.1 |
XM_017027923.1 |
1196 |
Missense Mutation |
ATG,GTG |
M147V |
XP_016883412.1 |
XM_017027924.1 |
1196 |
Missense Mutation |
ATG,GTG |
M115V |
XP_016883413.1 |
XM_017027925.1 |
1196 |
Missense Mutation |
ATG,GTG |
M91V |
XP_016883414.1 |
View Full Product Details