Product Details
- SNP ID
-
rs148426835
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:229296 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATACAGAATTTATTGGCTTGAGAC[A/G]CTGTTTAATGGGTTTGGGGAGATGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB129
PubMed Links
Gene Details
- Gene
- DEFB129
- Gene Name
- defensin beta 129
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080831.3 |
108 |
Missense Mutation |
CAC,CGC |
H26R |
NP_543021.1 |
View Full Product Details