Product Details

SNP ID
rs148702260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36614335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTTTCCAGTTGAGTGGTGTC[C/T]TCTGCACAGTCACAGGTAGGGGTAT
Phenotype
MIM: 606577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf24 PubMed Links

Gene Details

Gene
C20orf24
Gene Name
chromosome 20 open reading frame 24
There are no transcripts associated with this gene.

Gene
SLA2
Gene Name
Src like adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032214.3 1008 Missense Mutation AAG,AGG K212R NP_115590.1
NM_175077.2 1008 Silent Mutation GAA,GAG E195E NP_778252.1
XM_017028098.1 1008 Intron XP_016883587.1
Gene
TGIF2-C20orf24
Gene Name
TGIF2-C20orf24 readthrough
There are no transcripts associated with this gene.

View Full Product Details