Product Details

SNP ID
rs150011545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50128131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGATCTGGTTGGTGAAGGTGC[C/T]GAAGATGATCAGGCAGAAGACGAAG
Phenotype
MIM: 610994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM189 PubMed Links

Gene Details

Gene
TMEM189
Gene Name
transmembrane protein 189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162505.1 696 Missense Mutation AGC,GGC S176G NP_001155977.1
NM_199129.2 696 Missense Mutation AGC,GGC S179G NP_954580.1
Gene
TMEM189-UBE2V1
Gene Name
TMEM189-UBE2V1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199203.2 696 Missense Mutation AGC,GGC S179G NP_954673.1

View Full Product Details