Product Details

SNP ID
rs150059207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56634235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCGGCCGGGAGGCGGAAAGCC[A/G]CTCATGTGACTCTCCTGACATCCTT
Phenotype
MIM: 601602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFAP2C PubMed Links

Gene Details

Gene
TFAP2C
Gene Name
transcription factor AP-2 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003222.3 1132 Missense Mutation ACT,GCT T297A NP_003213.1

View Full Product Details