Product Details

SNP ID
rs150381386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5302190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACTTCATGGTGTTGTTCTTGAC[C/T]GTCACGAAGCACACGGTGTTGATCA
Phenotype
MIM: 607123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROKR2 PubMed Links

Gene Details

Gene
PROKR2
Gene Name
prokineticin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144773.3 1252 Silent Mutation NP_658986.1
XM_011529159.2 1252 Silent Mutation XP_011527461.1
XM_017027646.1 1252 Silent Mutation XP_016883135.1

View Full Product Details