Product Details

SNP ID
rs138816285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32731365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAACAGGAAACTTGTGATAGATA[A/G]AAATGAAGTATTTTCATTTGGAACA
Phenotype
MIM: 604297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAXBP1 PubMed Links

Gene Details

Gene
PAXBP1
Gene Name
PAX3 and PAX7 binding protein 1
There are no transcripts associated with this gene.

Gene
PAXBP1-AS1
Gene Name
PAXBP1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SYNJ1
Gene Name
synaptojanin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160302.1 Intron NP_001153774.1
NM_001160306.1 Intron NP_001153778.1
NM_003895.3 Intron NP_003886.3
NM_203446.2 Intron NP_982271.2

View Full Product Details