Product Details

SNP ID
rs138845591
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33581443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGTAAAGGTGTTGATTCCTTT[C/T]ACCAACACAACAGATTCAGGTCTGT
Phenotype
MIM: 603920 MIM: 611428 MIM: 182465
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CRYZL1 PubMed Links

Gene Details

Gene
CRYZL1
Gene Name
crystallin zeta like 1
There are no transcripts associated with this gene.

Gene
DONSON
Gene Name
downstream neighbor of SON
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017613.3 1276 Silent Mutation GTA,GTG V403V NP_060083.1
Gene
SON
Gene Name
SON DNA binding protein
There are no transcripts associated with this gene.

View Full Product Details