Product Details
- SNP ID
-
rs139598814
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:32576788 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTCCATCTTCAGACACCCCCCC[A/G]TCTCTCTGCACAGGGAGTTGGCCTT
- Phenotype
-
MIM: 608365
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCP10L
PubMed Links
Gene Details
- Gene
- TCP10L
- Gene Name
- t-complex 10-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144659.5 |
750 |
Missense Mutation |
CGG,TGG |
R212W |
NP_653260.1 |
View Full Product Details