Product Details

SNP ID
rs141886699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42472829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCACTTCATCTTAGTCCTGGA[A/G]GTCTGACTGTCTAGTTTCTTCTTCT
Phenotype
MIM: 609314
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RSPH1 PubMed Links

Gene Details

Gene
RSPH1
Gene Name
radial spoke head 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286506.1 1087 Missense Mutation CTC,TTC L269F NP_001273435.1
NM_080860.3 1087 Missense Mutation CTC,TTC L307F NP_543136.1
XM_005261208.2 1087 Missense Mutation CTC,TTC L238F XP_005261265.1
XM_011529786.1 1087 Missense Mutation CTC,TTC L283F XP_011528088.1

View Full Product Details