Product Details

SNP ID
rs144254488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44770996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCCCCATTTCTTCAGAAGGAG[C/T]GTTCTGGGTATTCCATCGTCCCCTG
Phenotype
MIM: 603124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2G2 PubMed Links

Gene Details

Gene
UBE2G2
Gene Name
ubiquitin conjugating enzyme E2 G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202489.1 885 UTR 3 NP_001189418.1
NM_003343.5 885 UTR 3 NP_003334.2
NM_182688.2 885 UTR 3 NP_872630.1
XM_017028469.1 885 UTR 3 XP_016883958.1

View Full Product Details