Product Details

SNP ID
rs144884596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33591207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCATCACATCCTTTAAGATA[C/T]GTAGCTGGAAGGATTGTTAAGGTGG
Phenotype
MIM: 603920 MIM: 611428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYZL1 PubMed Links

Gene Details

Gene
CRYZL1
Gene Name
crystallin zeta like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145858.2 1085 Missense Mutation TAT,TGT Y302C NP_665857.2
Gene
DONSON
Gene Name
downstream neighbor of SON
There are no transcripts associated with this gene.

View Full Product Details