Product Details

SNP ID
rs146322928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41171093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACTTCCTTCCCTTCATGCCTCCT[A/G]ATCTCTTTGGTTTTGGCCTCAAACT
Phenotype
MIM: 605668 MIM: 613770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BACE2 PubMed Links

Gene Details

Gene
BACE2
Gene Name
beta-site APP-cleaving enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012105.4 Intron NP_036237.2
NM_138991.2 Intron NP_620476.1
NM_138992.2 Intron NP_620477.1
XM_017028314.1 Intron XP_016883803.1
Gene
MIR3197
Gene Name
microRNA 3197
There are no transcripts associated with this gene.

Gene
PLAC4
Gene Name
placenta specific 4
There are no transcripts associated with this gene.

View Full Product Details