Product Details

SNP ID
rs146589453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42473108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATGCTAAGACAAGACTATCCA[G/T]GTAACATCAGTAATTCTCTTATGCC
Phenotype
MIM: 609314
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RSPH1 PubMed Links

Gene Details

Gene
RSPH1
Gene Name
radial spoke head 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286506.1 Intron NP_001273435.1
NM_080860.3 Intron NP_543136.1
XM_005261208.2 Intron XP_005261265.1
XM_011529786.1 Intron XP_011528088.1

View Full Product Details