Product Details

SNP ID
rs147370832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30747005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAAGTGGTCGGTAGCCACAGCA[A/G]CTAGAGCTGTATCCACAGCCATATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP21-1 PubMed Links

Gene Details

Gene
KRTAP21-1
Gene Name
keratin associated protein 21-1
There are no transcripts associated with this gene.

Gene
KRTAP21-2
Gene Name
keratin associated protein 21-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181617.1 198 Silent Mutation AGC,AGT S66S NP_853648.1

View Full Product Details