Product Details

SNP ID
rs147603846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30561168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAGCTGTATCTGTAGCCTCCA[G/T]AGCCACAGCCATAGCCCAGTCTGCG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP19-7 PubMed Links

Gene Details

Gene
KRTAP19-7
Gene Name
keratin associated protein 19-7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181614.1 122 Missense Mutation TAT,TCT Y41S NP_853645.1

View Full Product Details