Product Details

SNP ID
rs147639255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30282645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGACTTGGCCTGCTGATGGGGA[A/G]TTGCAGGGCACAGAGGAGTTTGACG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP24-1 PubMed Links

Gene Details

Gene
KRTAP24-1
Gene Name
keratin associated protein 24-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085455.2 314 Silent Mutation AAC,AAT N96N NP_001078924.1
Gene
KRTAP25-1
Gene Name
keratin associated protein 25-1
There are no transcripts associated with this gene.

View Full Product Details