Product Details

SNP ID
rs148054755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32794046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGCTTGGCCTCCATGTTGATGC[G/T]CAGCCTCTTCAGACCACAAATAGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C21orf62 PubMed Links

Gene Details

Gene
C21orf62
Gene Name
chromosome 21 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162495.2 673 Missense Mutation NP_001155967.2
NM_001162496.2 673 Missense Mutation NP_001155968.2
NM_019596.5 673 Missense Mutation NP_062542.5
XM_011529653.2 673 Intron XP_011527955.1
Gene
C21orf62-AS1
Gene Name
C21orf62 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details