Product Details

SNP ID
rs150222104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30430587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATGGATCTAGGTGCTGCTACTC[A/G]CTGGGAAATGGATCCAGTGGCTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP13-3 PubMed Links

Gene Details

Gene
KRTAP13-3
Gene Name
keratin associated protein 13-3
There are no transcripts associated with this gene.

Gene
KRTAP13-4
Gene Name
keratin associated protein 13-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181600.1 312 Silent Mutation TCA,TCG S104S NP_853631.1
Gene
KRTAP15-1
Gene Name
keratin associated protein 15-1
There are no transcripts associated with this gene.

View Full Product Details