Product Details

SNP ID
rs138882903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20917987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGCTCCGCCTGGTATATGGGGC[C/G]GGTGTCTCGCCAGGAGGCGCAGACC
Phenotype
MIM: 602007
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRKL PubMed Links

Gene Details

Gene
CRKL
Gene Name
CRK like proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005207.3 562 Missense Mutation CCG,CGG P18R NP_005198.1

View Full Product Details