Product Details

SNP ID
rs139791094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20982524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACTTCGGGCCCTTCGAAACAGT[G/T]CATCGCTGGCGGCGCCTCCCGCCCT
Phenotype
MIM: 600574
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AIFM3 PubMed Links

Gene Details

Gene
AIFM3
Gene Name
apoptosis inducing factor, mitochondria associated 3
There are no transcripts associated with this gene.

Gene
LZTR1
Gene Name
leucine zipper like transcription regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006767.3 256 Silent Mutation GTG,GTT V51V NP_006758.2

View Full Product Details