Product Details
- SNP ID
-
rs140214029
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:30370546 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGGAGTGTCCACTCCCAGACCA[C/T]TGAGACGGCCCTGGTGCCCTGTGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC157
PubMed Links
Gene Details
- Gene
- CCDC157
- Gene Name
- coiled-coil domain containing 157
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001017437.3 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
NP_001017437.2 |
NM_001318334.1 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
NP_001305263.1 |
NM_001318335.1 |
964 |
Intron |
|
|
NP_001305264.1 |
XM_006724278.3 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_006724341.1 |
XM_006724279.3 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_006724342.1 |
XM_006724280.3 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_006724343.1 |
XM_011530263.2 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_011528565.1 |
XM_011530266.2 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_011528568.1 |
XM_011530267.2 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_011528569.1 |
XM_011530268.2 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_011528570.1 |
XM_011530269.2 |
964 |
Intron |
|
|
XP_011528571.1 |
XM_011530270.2 |
964 |
Missense Mutation |
ACT,ATT |
T214I |
XP_011528572.1 |
- Gene
- KIAA1656
- Gene Name
- KIAA1656 protein
There are no transcripts associated with this gene.
- Gene
- RNF215
- Gene Name
- ring finger protein 215
There are no transcripts associated with this gene.
View Full Product Details