Product Details

SNP ID
rs140860349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37851477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAAGGTGTATGAGATCCAGGAC[A/G]TCTATGAGAACAGGTATGGGCTACT
Phenotype
MIM: 613383 MIM: 613556
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD54 PubMed Links

Gene Details

Gene
ANKRD54
Gene Name
ankyrin repeat domain 54
There are no transcripts associated with this gene.

Gene
EIF3L
Gene Name
eukaryotic translation initiation factor 3 subunit L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242923.1 358 Missense Mutation ATC,GTC I94V NP_001229852.1
NM_016091.3 358 Missense Mutation ATC,GTC I94V NP_057175.1
XM_005261625.1 358 Intron XP_005261682.1
XM_006724260.3 358 Missense Mutation ATC,GTC I137V XP_006724323.1
XM_017028814.1 358 Intron XP_016884303.1
Gene
MIR658
Gene Name
microRNA 658
There are no transcripts associated with this gene.

Gene
MIR659
Gene Name
microRNA 659
There are no transcripts associated with this gene.

View Full Product Details