Product Details

SNP ID
rs141244105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20425956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGCGCACGGCCAGCTGCAGCGG[C/T]GCTGTGCTTGCCGTGGATCCAGGAC
Phenotype
MIM: 613619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARF2 PubMed Links

Gene Details

Gene
SCARF2
Gene Name
scavenger receptor class F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153334.6 2125 Missense Mutation ACC,GCC T679A NP_699165.3
NM_182895.4 2125 Missense Mutation ACC,GCC T674A NP_878315.2
XM_017029065.1 2125 Intron XP_016884554.1

View Full Product Details