Product Details

SNP ID
rs142196057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21958507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACTCACCTTTCTTCATGTCTC[A/G]GAAGGGTGGGTGGTTGTAGCAGTAG
Phenotype
MIM: 603582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPM1F PubMed Links

Gene Details

Gene
PPM1F
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1F
There are no transcripts associated with this gene.

Gene
TOP3B
Gene Name
topoisomerase (DNA) III beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282112.1 2345 Missense Mutation NP_001269041.1
NM_001282113.1 2345 Missense Mutation NP_001269042.1
NM_003935.4 2345 Missense Mutation NP_003926.1
XM_005261811.1 2345 Missense Mutation XP_005261868.1
XM_005261813.2 2345 Nonsense Mutation XP_005261870.1
XM_006724349.1 2345 Nonsense Mutation XP_006724412.1
XM_006724350.1 2345 Nonsense Mutation XP_006724413.1
XM_011530482.1 2345 Nonsense Mutation XP_011528784.1
XM_017029038.1 2345 Nonsense Mutation XP_016884527.1
XM_017029039.1 2345 Nonsense Mutation XP_016884528.1
XM_017029040.1 2345 Nonsense Mutation XP_016884529.1
XM_017029041.1 2345 Nonsense Mutation XP_016884530.1

View Full Product Details