Product Details

SNP ID
rs142559213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30928137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTCCTCGGAGGCCTTGGCCCGG[C/G]AGTCAGCACGGCTCTGGTAGGAATT
Phenotype
MIM: 616661
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MORC2 PubMed Links

Gene Details

Gene
MORC2
Gene Name
MORC family CW-type zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303256.2 3310 Missense Mutation TCC,TGC S971C NP_001290185.1
NM_001303257.2 3310 Missense Mutation TCC,TGC S971C NP_001290186.1
NM_014941.3 3310 Missense Mutation TCC,TGC S909C NP_055756.1
XM_011530004.1 3310 Missense Mutation TCC,TGC S976C XP_011528306.1
XM_017028667.1 3310 Missense Mutation TCC,TGC S976C XP_016884156.1
XM_017028668.1 3310 Missense Mutation TCC,TGC S909C XP_016884157.1
Gene
MORC2-AS1
Gene Name
MORC2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details