Product Details

SNP ID
rs144622988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37850041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTTATGACCCCTACGCTTATCC[C/T]AGCGACTATGATATGCACACAGGTG
Phenotype
MIM: 613383 MIM: 613556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD54 PubMed Links

Gene Details

Gene
ANKRD54
Gene Name
ankyrin repeat domain 54
There are no transcripts associated with this gene.

Gene
EIF3L
Gene Name
eukaryotic translation initiation factor 3 subunit L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242923.1 138 Silent Mutation CCC,CCT P20P NP_001229852.1
NM_016091.3 138 Silent Mutation CCC,CCT P20P NP_057175.1
XM_005261625.1 138 Intron XP_005261682.1
XM_006724260.3 138 Silent Mutation CCC,CCT P63P XP_006724323.1
XM_017028814.1 138 Intron XP_016884303.1
Gene
MIR658
Gene Name
microRNA 658
There are no transcripts associated with this gene.

Gene
MIR659
Gene Name
microRNA 659
There are no transcripts associated with this gene.

View Full Product Details