Product Details

SNP ID
rs144674196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39081125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTACCCTGACCATCTTTGTTGCC[C/T]GCCTCTACTACTTCTGGGACCCAGA
Phenotype
MIM: 607113
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOBEC3G PubMed Links

Gene Details

Gene
APOBEC3G
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021822.3 721 Missense Mutation CGC,TGC R122C NP_068594.1
XM_006724290.1 721 Missense Mutation CGC,TGC R55C XP_006724353.1
XM_017028903.1 721 Missense Mutation CGC,TGC R122C XP_016884392.1
XM_017028904.1 721 Missense Mutation CGC,TGC R55C XP_016884393.1

View Full Product Details