Product Details

SNP ID
rs146323944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20918042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGGCCAGCGCCACGGTATGTT[C/T]CTCGTCCGCGATTCTTCCACCTGCC
Phenotype
MIM: 602007
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRKL PubMed Links

Gene Details

Gene
CRKL
Gene Name
CRK like proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005207.3 617 Silent Mutation TTC,TTT F36F NP_005198.1

View Full Product Details