Product Details

SNP ID
rs147334255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27750995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGCCAGGTGGGCACGGAGGCTC[A/G]AGCCTTGGCGTCACCCACGTCGTCT
Phenotype
MIM: 156100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MN1 PubMed Links

Gene Details

Gene
MN1
Gene Name
MN1 proto-oncogene, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002430.2 4838 Nonsense Mutation CGA,TGA R1295* NP_002421.3

View Full Product Details