Product Details

SNP ID
rs147963916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31934369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGATTGAAGAAAGGGAGAAATCC[A/G]GCTCTGTGGGCTAGGGCAGGTGCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf24 PubMed Links

Gene Details

Gene
C22orf24
Gene Name
chromosome 22 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302819.1 411 Missense Mutation CGG,TGG R145W NP_001289748.1
NM_001302820.1 411 UTR 3 NP_001289749.1
NM_015372.2 411 Missense Mutation CGG,TGG R78W NP_056187.1

View Full Product Details