Product Details

SNP ID
rs148160537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50061368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTTACGACACGGGAGCCACTC[A/G]GAGCTGACTGATCTCACTGAGGCAC
Phenotype
MIM: 605908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MLC1 PubMed Links

Gene Details

Gene
MLC1
Gene Name
megalencephalic leukoencephalopathy with subcortical cysts 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015166.3 1579 UTR 3 NP_055981.1
NM_139202.2 1579 UTR 3 NP_631941.1
XM_011530678.1 1579 UTR 3 XP_011528980.1
XM_017028671.1 1579 UTR 3 XP_016884160.1
Gene
TTLL8
Gene Name
tubulin tyrosine ligase like 8
There are no transcripts associated with this gene.

View Full Product Details